Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014167.4 | NP_054886.2 | 2 | 252 | ||
NR_033192.1 | 2 | 181 | ||||
GenBank mRNA | AF161477.1 | - | 2 | 226 | ||
AF213377.1 | - | 2 | 219 | |||
AK001156.1 | - | 2 | 228 | |||
AK225139.1 | - | 2 | 231 | |||
BC020647.1 | - | 2 | 215 | 4771954 | ||
BG538660.1 | - | 2 | 257 | |||
BM846580.1 | - | 2 | 151 | |||
DA666239.1 | - | 2 | 181 |