Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293186.1 | NP_001280115.1 | 4 | 437 | ||
NM_001293187.1 | NP_001280116.1 | 4 | 351 | |||
NM_014168.3 | NP_054887.2 | 3 | 578 | |||
XM_011511023.1 | XP_011509325.1 | 4 | 378 | |||
GenBank mRNA | AF161482.1 | - | 4 | 326 | ||
AF201938.1 | - | 4 | 395 | |||
AK001321.1 | - | 4 | 325 | |||
AL523293.3 | - | 3 | 560 | |||
BC000921.2 | - | 3 | 490 | 3445582 | ||
BC093014.1 | - | 4 | 333 | 4799279 | ||
CA337169.1 | - | 4 | 352 |