Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTPBP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014170.2 | NP_054889.2 | 4 | 699 | ||
NM_138485.1 | NP_612494.1 | 3 | 600 | |||
GenBank mRNA | AF161484.1 | - | 4 | 738 | ||
AK093419.1 | - | 4 | 1293 | |||
AK289611.1 | - | 3 | 562 | |||
AK292651.1 | - | 4 | 718 | |||
BC010137.2 | - | 2 | 435 | |||
BC037163.1 | - | 4 | 702 | 4620206 | ||
BG655089.1 | - | 3 | 502 | |||
BI830308.1 | - | 4 | 699 | |||
BX354218.2 | - | 3 | 568 | |||
CK818022.1 | - | 3 | 489 |