Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTPBP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014170.2 | NP_054889.2 | 5 | 792 | ||
NM_138485.1 | NP_612494.1 | 4 | 693 | |||
GenBank mRNA | AF161484.1 | - | 5 | 831 | ||
AK093419.1 | - | 5 | 1386 | |||
AK289611.1 | - | 4 | 655 | |||
AK292651.1 | - | 5 | 811 | |||
BC010137.2 | - | 3 | 528 | |||
BC037163.1 | - | 5 | 795 | 4620206 | ||
BX354218.2 | - | 4 | 661 |