Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135861.2 | NP_001129333.1 | 2 | 869 | ||
NM_001287342.1 | NP_001274271.1 | 2 | 875 | |||
NM_001287343.1 | NP_001274272.1 | 2 | 869 | |||
NM_014172.5 | NP_054891.2 | 2 | 869 | |||
NR_109807.1 | 2 | 869 | ||||
NR_109808.1 | 2 | 852 | ||||
GenBank mRNA | AF131857.1 | - | 2 | 593 | ||
AF161490.1 | - | 2 | 274 | |||
AF164795.1 | - | 2 | 262 | |||
AF285119.1 | - | 2 | 587 | |||
AF393504.1 | - | 2 | 254 | |||
AL136644.1 | - | 2 | 279 | |||
BC024648.1 | - | 2 | 275 | 4934238 | ||
BP375594.1 | - | 2 | 273 | |||
BQ922335.1 | - | 2 | 284 | |||
CD242442.1 | - | 2 | 482 | |||
CD693554.1 | - | 2 | 314 | |||
CN334799.1 | - | 2 | 252 | |||
CR533481.1 | - | 2 | 254 | |||
EU794646.1 | - | 2 | 586 | |||
F29783.1 | - | 2 | 112 |