Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2T | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310326.1 | NP_001297255.1 | 3 | 292 | ||
NM_014176.3 | NP_054895.1 | 3 | 288 | |||
GenBank mRNA | AB032931.1 | - | 2 | 194 | ||
AF160215.1 | - | 3 | 245 | |||
AF161499.1 | - | 3 | 278 | |||
AK000504.1 | - | 3 | 243 | |||
AY542309.1 | - | 2 | 153 | |||
BC004152.2 | - | 3 | 250 | 2821806 | ||
BC019284.1 | - | 3 | 250 | 2821806 | ||
BG776416.1 | - | 3 | 289 | |||
BM460257.1 | - | 3 | 265 | |||
BP244265.1 | - | 3 | 288 |