Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014177.2 | NP_054896.2 | 6 | 955 | ||
XM_005266689.3 | XP_005266746.1 | 6 | 956 | |||
GenBank mRNA | AF161503.1 | - | 6 | 856 | ||
AK289923.1 | - | 6 | 953 | |||
AK313936.1 | - | 6 | 943 | |||
BC000892.1 | - | 6 | 915 | 3463640 |