Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STXBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304476.1 | NP_001291405.1 | 5, 6 | 1013 | ||
NM_001304477.1 | NP_001291406.1 | 4, 5 | 518 | |||
NM_014178.7 | NP_054897.4 | 4, 5 | 1160 | |||
XM_006720121.3 | XP_006720184.1 | 4, 5 | 523 | |||
XM_011536669.2 | XP_011534971.1 | 3, 4 | 555 | |||
XM_017021231.1 | XP_016876720.1 | 4, 5 | 892 | |||
XM_017021232.1 | XP_016876721.1 | 5, 6 | 3488 | |||
XM_017021233.1 | XP_016876722.1 | 5, 6 | 840 | |||
XM_017021234.1 | XP_016876723.1 | 5, 6 | 613 | |||
XM_017021235.1 | XP_016876724.1 | 4, 5 | 3401 | |||
XM_017021236.1 | XP_016876725.1 | 5, 6 | 817 | |||
XM_017021237.1 | XP_016876726.1 | 5, 6 | 646 | |||
XM_017021238.1 | XP_016876727.1 | 4, 5 | 584 | |||
XM_017021239.1 | XP_016876728.1 | 4, 5 | 582 | |||
XM_017021240.1 | XP_016876729.1 | 4, 5 | 504 | |||
XM_017021241.1 | XP_016876730.1 | 4, 5 | 1121 | |||
XM_017021242.1 | XP_016876731.1 | 5, 6 | 586 | |||
GenBank mRNA | AF391153.1 | - | 3, 4 | 484 | ||
AK096957.1 | - | 6, 7 | 743 | |||
AK309203.1 | - | 4, 5 | 518 | |||
AL834346.1 | - | 4, 5 | 498 | |||
BC009499.2 | - | 5, 6 | 787 | 3955370 | ||
BC067278.1 | - | 4, 5 | 1160 | 6178786 |