Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM208 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318217.1 | NP_001305146.1 | 3 | 354 | ||
NM_014187.3 | NP_054906.2 | 3 | 237 | |||
NR_134524.1 | 3 | 237 | ||||
GenBank mRNA | AF161519.1 | - | 3 | 239 | ||
BC003080.2 | - | 3 | 210 | 3050277 | ||
BC013412.2 | - | 3 | 189 | 3030307 | ||
BC021109.1 | - | 3 | 193 | 3660106 | ||
BF316569.1 | - | 3 | 164 | |||
BG825054.1 | - | 3 | 315 | |||
BI833541.1 | - | 3 | 196 | |||
BI838179.1 | - | 2 | 660 |