Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM208 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318217.1 | NP_001305146.1 | 5 | 534 | ||
NM_014187.3 | NP_054906.2 | 5 | 417 | |||
NR_134524.1 | 5 | 443 | ||||
GenBank mRNA | AF161519.1 | - | 5 | 440 | ||
BC003080.2 | - | 5 | 390 | 3050277 | ||
BC013412.2 | - | 5 | 369 | 3030307 | ||
BC021109.1 | - | 5 | 399 | 3660106 | ||
BF316569.1 | - | 5 | 343 | |||
BG825054.1 | - | 5 | 495 | |||
BI833541.1 | - | 5 | 402 |