Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173984.2 | NP_001167455.1 | 2 | 321 | ||
NM_013263.4 | NP_037395.2 | 2 | 321 | |||
XM_011523046.2 | XP_011521348.1 | 2 | 356 | |||
XM_011523047.2 | XP_011521349.1 | 2 | 356 | |||
XM_011523048.2 | XP_011521350.1 | 2 | 356 | |||
XM_011523049.2 | XP_011521351.1 | 2 | 356 | |||
XM_017023179.1 | XP_016878668.1 | 2 | 221 | |||
XM_017023180.1 | XP_016878669.1 | 2 | 221 | |||
GenBank mRNA | AF152604.1 | - | 2 | 214 | ||
AF213969.1 | - | 2 | 166 | |||
AJ271881.1 | - | 2 | 279 | |||
BC050728.1 | - | 3 | 228 | 6149862 | ||
BC094706.1 | - | 3 | 198 | 30719813 | ||
DA645362.1 | - | 2 | 221 |