Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013264.4 | NP_037396.3 | 10 | 1224 | ||
XM_011542790.2 | XP_011541092.1 | 10 | 1128 | |||
XM_011542791.2 | XP_011541093.1 | 10 | 1176 | |||
XM_017017626.1 | XP_016873115.1 | 10 | 1165 | |||
GenBank mRNA | AF155140.2 | - | 10 | 1118 | ||
AK295121.1 | - | 10 | 1084 | |||
AK312772.1 | - | 10 | 1085 | |||
BC035388.1 | - | 10 | 1070 | 5166189 | ||
BC050360.1 | - | 10 | 1094 | 5742404 | ||
CD657404.1 | - | 2 | 169 |