Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD274 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001314029.1 | NP_001300958.1 | 2, 3 | 154 | ||
NM_014143.3 | NP_054862.1 | 2, 3 | 154 | |||
NR_052005.1 | 2, 3 | 154 | ||||
GenBank mRNA | AF177937.1 | - | 1, 2 | 46 | ||
AF233516.1 | - | 2, 3 | 98 | |||
AK300470.1 | - | 2, 3 | 115 | |||
AK314567.1 | - | 2, 3 | 115 | |||
AY254342.1 | - | 1, 2 | 46 | |||
AY714881.1 | - | 1, 2 | 46 | |||
BC069381.1 | - | 2, 3 | 96 | 7262208 | ||
BC074984.2 | - | 1, 2 | 66 | 30915301 | ||
BC113734.1 | - | 1, 2 | 67 | 8322786 | ||
BC113736.1 | - | 1, 2 | 67 | 8322788 | ||
DB160805.1 | - | 2, 3 | 85 | |||
DQ836393.1 | - | 2, 3 | 159 |