Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD274 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267706.1 | NP_001254635.1 | 3 | 400 | ||
NM_001314029.1 | NP_001300958.1 | 4 | 742 | |||
NM_014143.3 | NP_054862.1 | 4 | 742 | |||
GenBank mRNA | AF177937.1 | - | 3 | 634 | ||
AF233516.1 | - | 4 | 686 | |||
AK001894.1 | - | 2 | 449 | |||
AK314567.1 | - | 4 | 703 | |||
AY254342.1 | - | 3 | 634 | |||
AY291313.1 | - | 2 | 292 | |||
BC069381.1 | - | 4 | 684 | 7262208 | ||
BC074984.2 | - | 3 | 654 | 30915301 | ||
BC113734.1 | - | 3 | 655 | 8322786 | ||
BC113736.1 | - | 3 | 655 | 8322788 | ||
DQ286582.1 | - | 2 | 292 |