Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RACGAP1 SIRNA Assays ›
Gene Symbol:
RACGAP1
Gene Name:
Rac GTPase activating protein 1
Gene Aliases:
CYK4, HsCYK-4, ID-GAP, MgcRacGAP
Chromosome Location:
Chr.12: 49989162 - 50033136 on Build GRCh38
UniGene ID:
Hs.505469
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s26550
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RACGAP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001126103.2
NP_001119575.1
10
1021
NM_001126104.2
NP_001119576.1
8
901
NM_001319999.1
NP_001306928.1
8
831
NM_001320000.1
NP_001306929.1
9
905
NM_001320001.1
NP_001306930.1
8
897
NM_001320002.1
NP_001306931.1
8
856
NM_001320003.1
NP_001306932.1
8
827
NM_001320004.1
NP_001306933.1
8
778
NM_001320005.1
NP_001306934.1
7
742
NM_001320006.1
NP_001306935.1
9
901
NM_001320007.1
NP_001306936.1
7
628
NM_013277.4
NP_037409.2
10
1010
XM_006719359.1
XP_006719422.1
8
729
XM_011538238.1
XP_011536540.1
8
686
XM_011538243.1
XP_011536545.1
8
806
XM_017019220.1
XP_016874709.1
8
1003
XM_017019221.1
XP_016874710.1
7
647
XM_017019222.1
XP_016874711.1
7
602
XM_017019223.1
XP_016874712.1
7
643
XM_017019224.1
XP_016874713.1
7
573
XM_017019225.1
XP_016874714.1
7
482
XM_017019226.1
XP_016874715.1
7
542
XM_017019227.1
XP_016874716.1
7
563
GenBank mRNA
AB030251.1
-
8
737
AB040911.1
-
6
482
AK000733.1
-
8
744
AK225901.1
-
7
672
AK315785.1
-
10
958
AL136794.1
-
10
891
BC024144.2
-
8
722
BC032754.1
-
8
728
5583315
CR533565.1
-
7
667
DA500396.1
-
7
548
Back To Top
More Information
Panther Classification:
Molecular Function -
GTPase-activating protein
Gene Ontology Categories:
Function(s)
Process(es)
mitotic cytokinesis
actomyosin contractile ring assembly
retrograde vesicle-mediated transport, Golgi to ER
microtubule-based movement
spermatogenesis
neuroblast proliferation
sulfate transport
embryo development
antigen processing and presentation of exogenous peptide antigen via MHC class II
positive regulation of cytokinesis
intracellular signal transduction
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
mitotic spindle midzone assembly
regulation of attachment of spindle microtubules to kinetochore
GTPase activator activity
protein binding
phosphatidylinositol-3,4,5-trisphosphate binding
microtubule binding
protein kinase binding
alpha-tubulin binding
gamma-tubulin binding
metal ion binding
beta-tubulin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-xbcvc:80/10.239.164.135:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline