Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UHRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048201.2 | NP_001041666.1 | 11 | 1726 | ||
NM_001290050.1 | NP_001276979.1 | 11 | 2066 | |||
NM_001290051.1 | NP_001276980.1 | 11 | 1654 | |||
NM_001290052.1 | NP_001276981.1 | 11 | 1628 | |||
NM_013282.4 | NP_037414.3 | 10 | 2009 | |||
XM_011527942.2 | XP_011526244.1 | 10 | 1550 | |||
GenBank mRNA | AB177623.1 | - | 11 | 2066 | ||
AB177624.1 | - | 10 | 2009 | |||
AF129507.1 | - | 10 | 1490 | |||
AF274048.1 | - | 10 | 1574 | |||
AK025578.1 | - | 1 | 56 | |||
AK074377.1 | - | 11 | 1655 | |||
AK289389.1 | - | 11 | 1567 | |||
AK303361.1 | - | 6 | 599 | |||
AK314579.1 | - | 11 | 1655 | |||
BC113875.1 | - | 10 | 1574 | 40053955 | ||
EF560733.1 | - | 11 | 1621 |