Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMOD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014547.4 | NP_055362.1 | 5 | 712 | ||
XM_017022087.1 | XP_016877576.1 | 6 | 743 | |||
XM_017022088.1 | XP_016877577.1 | 5 | 584 | |||
GenBank mRNA | AF177171.1 | - | 5 | 674 | ||
AF237631.1 | - | 4 | 494 | |||
AK310344.1 | - | 3 | 391 | |||
AK312569.1 | - | 5 | 709 | |||
BC020542.2 | - | 5 | 648 | 4421113 | ||
BX647277.1 | - | 5 | 674 |