Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014550.3 | NP_055365.2 | 14 | 2297 | ||
GenBank mRNA | AK074906.1 | - | 11 | 1527 | ||
AK296176.1 | - | 2 | 567 | |||
AY028896.1 | - | 14 | 2334 | |||
AY032928.1 | - | 14 | 2297 | |||
BC032058.1 | - | 12 | 1922 | |||
BC113659.1 | - | 14 | 2315 | 8322711 | ||
BC117403.1 | - | 14 | 2309 | 40125954 |