Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCAPH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185011.1 | NP_001171940.1 | 5 | 513 | ||
NM_014551.4 | NP_055366.3 | 5 | 513 | |||
NM_152299.3 | NP_689512.2 | 5 | 513 | |||
XM_005261912.4 | XP_005261969.1 | 6 | 565 | |||
XM_011530685.2 | XP_011528987.1 | 5 | 393 | |||
XM_017028793.1 | XP_016884282.1 | 6 | 551 | |||
GenBank mRNA | AL021682.1 | - | 5 | 482 | ||
BC000473.2 | - | 5 | 486 | |||
BC001509.2 | - | 5 | 486 | |||
BC001833.2 | - | 5 | 486 | |||
BC001937.1 | - | 5 | 470 | 2819717 | ||
BC009441.1 | - | 5 | 469 | 3509636 | ||
BC014939.1 | - | 3 | 173 | |||
CR456604.1 | - | 5 | 445 |