Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198182.2 | NP_937825.2 | 6 | 929 | ||
XM_005246159.3 | XP_005246216.1 | 5 | 815 | |||
XM_006711882.2 | XP_006711945.1 | 6 | 929 | |||
XM_006711884.3 | XP_006711947.1 | 5 | 808 | |||
XM_011510343.2 | XP_011508645.1 | 6 | 900 | |||
XM_017003900.1 | XP_016859389.1 | 6 | 1002 | |||
XM_017003901.1 | XP_016859390.1 | 6 | 1002 | |||
GenBank mRNA | AF198489.1 | - | 6 | 867 | ||
AF411210.1 | - | 2 | 284 | |||
AK293855.1 | - | 6 | 976 | |||
AK296982.1 | - | 6 | 1007 | |||
AK312950.1 | - | 6 | 899 | |||
BC067519.1 | - | 6 | 810 | 6971723 | ||
BC067520.1 | - | 6 | 810 | 6971724 | ||
BC067521.1 | - | 6 | 810 | 6971725 |