Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321792.1 | NP_001308721.1 | 5 | 478 | ||
NM_013342.3 | NP_037474.1 | 5 | 872 | |||
XM_005278261.1 | XP_005278318.1 | 4 | 348 | |||
GenBank mRNA | AF052052.1 | - | 6 | 745 | ||
BC004281.1 | - | 5 | 504 | 3616596 | ||
BC007776.2 | - | 5 | 490 | 4301790 |