Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ICOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012092.3 | NP_036224.1 | 2 | 296 | ||
GenBank mRNA | AB023135.1 | - | 2 | 254 | ||
AJ277832.1 | - | 2 | 296 | |||
AK314985.1 | - | 2 | 313 | |||
BC028006.1 | - | 2 | 264 | 5212706 | ||
BC028210.1 | - | 2 | 264 | 5215237 |