Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142364.1 | NP_001135836.1 | 9 | 872 | ||
NM_013338.4 | NP_037470.1 | 10 | 962 | |||
XM_017020552.1 | XP_016876041.1 | 9 | 900 | |||
GenBank mRNA | AF088028.1 | - | 5 | 400 | ||
AF102850.1 | - | 10 | 970 | |||
AF161498.1 | - | 10 | 911 | |||
AF183413.1 | - | 10 | 921 | |||
AK299085.1 | - | 9 | 848 | |||
BC012531.1 | - | 10 | 914 | 4273297 |