Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM216A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013300.2 | NP_037432.2 | 4 | 809 | ||
XM_005253875.4 | XP_005253932.1 | 4 | 319 | |||
XM_011538246.2 | XP_011536548.1 | 4 | 337 | |||
GenBank mRNA | AK225731.1 | - | 4 | 800 | ||
AK296280.1 | - | 4 | 821 | |||
AK297684.1 | - | 4 | 319 | |||
AK308802.1 | - | 4 | 422 | |||
AK309603.1 | - | 3 | 333 | |||
BC020967.1 | - | 4 | 625 | 3848037 | ||
CR457001.1 | - | 4 | 360 | |||
U79274.1 | - | 4 | 776 |