Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NM_013301.2 | NP_037433.2 | 3 | 835 | ||
XM_005258827.2 | XP_005258884.1 | 3 | 433 | |||
XM_005258828.2 | XP_005258885.1 | 3 | 454 | |||
GenBank mRNA | AF054984.1 | - | 3 | 388 | ||
AK097095.1 | - | 2 | 1005 | |||
AY927563.1 | - | 1 | 370 | |||
AY927628.1 | - | LIT3614 AY927628 |
2 | 243 | ||
BC000412.2 | - | 2 | 413 | 2961290 | ||
BC008956.2 | - | 2 | 413 | 2961290 | ||
CR450308.1 | - | 2 | 100 | |||
GU354321.1 | - | 3 | 190 | |||
GU354322.1 | - | 3 | 246 | |||
GU354323.1 | - | 3 | 434 | |||
U79303.1 | - | 3 | 388 |