Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013306.4 | NP_037438.2 | 8 | 1082 | ||
NM_147777.3 | NP_680086.2 | 7 | 824 | |||
GenBank mRNA | AB209439.1 | - | 7 | 3860 | ||
AF001435.1 | - | 8 | 1011 | |||
AF175267.2 | - | 8 | 991 | |||
AF175268.1 | - | 7 | 691 | |||
AK315034.1 | - | 8 | 1013 | |||
AK315642.1 | - | 8 | 1023 | |||
BC009897.2 | - | 8 | 1000 | 3510379 | ||
BC012767.2 | - | 8 | 1052 | 3678551 | ||
BC014520.2 | - | 8 | 1052 | 3678576 | ||
BE547440.1 | - | 2 | 92 | |||
BE884806.1 | - | 2 | 158 | |||
BT006631.1 | - | 8 | 952 |
Gene Symbol: ARL2-SNX15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037650.1 | 11 | 1606 | |||
GenBank mRNA | AK096447.1 | - | 11 | 1540 |