Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCFC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013320.2 | NP_037452.1 | 9, 10 | 1383 | ||
XM_017019241.1 | XP_016874730.1 | 8, 9 | 1215 | |||
XM_017019242.1 | XP_016874731.1 | 9, 10 | 1383 | |||
GenBank mRNA | AF117210.1 | - | 9, 10 | 1292 | ||
AK223342.1 | - | 9, 10 | 1310 | |||
AK296523.1 | - | 2, 3 | 165 | |||
AK313466.1 | - | 9, 10 | 1313 | |||
BC033799.1 | - | 9, 10 | 1285 | 5172046 |