Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PYCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271681.1 | NP_001258610.1 | 1, 2 | 306 | ||
NM_013328.3 | NP_037460.2 | 1, 2 | 306 | |||
GenBank mRNA | AK001508.1 | - | 1, 2 | 244 | ||
AK291913.1 | - | 1, 2 | 141 | |||
BC014868.1 | - | 1, 2 | 197 | 3919161 | ||
BC020553.1 | - | 1, 2 | 231 | 2901360 | ||
BU538765.1 | - | 1, 2 | 181 | |||
HY044355.1 | - | 1, 2 | 307 | |||
KU178615.1 | - | 1, 2 | 76 | |||
KU178616.1 | - | 1, 2 | 76 |