Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130071.1 | NP_001123543.1 | 11 | 2896 | ||
NM_001130072.1 | NP_001123544.1 | 11 | 2433 | |||
NM_013333.3 | NP_037465.2 | 10 | 2355 | |||
XM_005258829.2 | XP_005258886.1 | 11 | 2442 | |||
XM_005258830.2 | XP_005258887.1 | 11 | 2430 | |||
XM_011526880.1 | XP_011525182.1 | 11 | 2439 | |||
XM_011526881.1 | XP_011525183.1 | 10 | 2367 | |||
XM_017026722.1 | XP_016882211.1 | 10 | 2364 | |||
XM_017026723.1 | XP_016882212.1 | 13 | 2484 | |||
XM_017026724.1 | XP_016882213.1 | 13 | 2422 | |||
XM_017026725.1 | XP_016882214.1 | 12 | 2409 | |||
GenBank mRNA | AK022454.1 | - | 12 | 2402 | ||
BC044651.1 | - | 10 | 2305 | 5244118 |