Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130071.1 | NP_001123543.1 | 3 | 924 | ||
NM_001130072.1 | NP_001123544.1 | 2 | 386 | |||
NM_013333.3 | NP_037465.2 | 2 | 386 | |||
XM_005258829.2 | XP_005258886.1 | 2 | 395 | |||
XM_005258830.2 | XP_005258887.1 | 2 | 386 | |||
XM_011526880.1 | XP_011525182.1 | 2 | 395 | |||
XM_011526881.1 | XP_011525183.1 | 2 | 395 | |||
XM_017026722.1 | XP_016882211.1 | 2 | 395 | |||
XM_017026723.1 | XP_016882212.1 | 4 | 437 | |||
XM_017026724.1 | XP_016882213.1 | 4 | 375 | |||
XM_017026725.1 | XP_016882214.1 | 4 | 437 | |||
GenBank mRNA | AF073727.1 | - | 3 | 923 | ||
AK022454.1 | - | 2 | 355 | |||
AK314690.1 | - | 2 | 356 | |||
BC044651.1 | - | 2 | 336 | 5244118 | ||
DA657018.1 | - | 2 | 387 |