Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC61A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013336.3 | NP_037468.1 | 10 | 1265 | ||
GenBank mRNA | AF084458.1 | - | 10 | 1171 | ||
AF346602.1 | - | 10 | 1147 | |||
AK001689.1 | - | 3 | 325 | |||
AK027423.1 | - | 10 | 1265 | |||
AK027452.1 | - | 2 | 231 | |||
AK027454.1 | - | 1 | 51 | |||
AK074706.1 | - | 10 | 1265 | |||
AK074907.1 | - | 2 | 235 | |||
AK074928.1 | - | 6 | 873 | |||
AK075148.1 | - | 2 | 265 | |||
AK299118.1 | - | 10 | 1345 | |||
AL831940.1 | - | 5 | 675 | |||
AL832821.1 | - | 10 | 1198 | |||
BC002951.1 | - | 10 | 1172 |