Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC61A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013336.3 | NP_037468.1 | 6 | 628 | ||
GenBank mRNA | AF077032.1 | - | 6 | 534 | ||
AF084458.1 | - | 6 | 534 | |||
AF346602.1 | - | 6 | 510 | |||
AK027423.1 | - | 6 | 628 | |||
AK074706.1 | - | 6 | 628 | |||
AK074928.1 | - | 2 | 236 | |||
AK299118.1 | - | 6 | 708 | |||
AL832821.1 | - | 6 | 561 | |||
BC002951.1 | - | 6 | 535 |