Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NENF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013349.4 | NP_037481.1 | 4 | 440 | ||
NR_026598.1 | 3 | 379 | ||||
GenBank mRNA | AB126219.1 | - | 4 | 383 | ||
AF049672.1 | - | 4 | 238 | |||
AF173937.1 | - | 4 | 443 | |||
AK074431.1 | - | 4 | 408 | |||
AK223135.1 | - | 4 | 408 | |||
AY762102.1 | - | 4 | 399 | |||
BC008823.2 | - | 4 | 389 | 4303614 | ||
DB502983.1 | - | 3 | 355 |