Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080976.2 | NP_001074445.1 | 2, 3 | 665 | ||
NM_001322937.1 | NP_001309866.1 | 3, 4 | 1221 | |||
NM_001322938.1 | NP_001309867.1 | 3, 4 | 1133 | |||
NM_001322939.1 | NP_001309868.1 | 2, 3 | 548 | |||
NM_001322943.1 | NP_001309872.1 | 2, 3 | 719 | |||
NM_001322944.1 | NP_001309873.1 | 2, 3 | 719 | |||
NM_013352.3 | NP_037484.1 | 2, 3 | 719 | |||
XM_017010795.1 | XP_016866284.1 | 4, 5 | 854 | |||
XM_017010796.1 | XP_016866285.1 | 1, 2 | 475 | |||
XM_017010797.1 | XP_016866286.1 | 2, 3 | 1399 | |||
GenBank mRNA | AF098066.1 | - | 2, 3 | 563 | ||
AK293872.1 | - | 1112 | ||||
AK301513.1 | - | 2, 3 | 548 | |||
BC039245.1 | - | 2, 3 | 610 | 5272885 |