Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANAPC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286756.1 | NP_001273685.1 | 3 | 284 | ||
NM_013367.2 | NP_037499.2 | 3 | 284 | |||
XM_011513838.1 | XP_011512140.1 | 3 | 195 | |||
XM_011513839.1 | XP_011512141.1 | 3 | 205 | |||
XM_017008138.1 | XP_016863627.1 | 3 | 205 | |||
GenBank mRNA | AF191338.1 | - | 2 | 155 | ||
AK292336.1 | - | 3 | 204 | |||
BC059383.1 | - | 3 | 257 | 30349524 | ||
BI546948.1 | - | 3 | 289 |