Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNMT3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013369.3 | NP_037501.2 | 2, 3 | 593 | ||
NM_175867.2 | NP_787063.1 | 2, 3 | 593 | |||
GenBank mRNA | AF194032.1 | - | 2, 3 | 593 | ||
BC002560.2 | - | 2, 3 | 267 | 3138514 |