Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013386.4 | NP_037518.3 | 5, 6 | 897 | ||
NM_213651.2 | NP_998816.1 | 5, 6 | 808 | |||
XM_017001104.1 | XP_016856593.1 | 1, 2 | 153 | |||
GenBank mRNA | AF123303.1 | - | 5, 6 | 656 | ||
AJ619961.1 | - | 5, 6 | 740 | |||
AJ619987.1 | - | 5, 6 | 686 | |||
AK292567.1 | - | 5, 6 | 808 | |||
AK304165.1 | - | 5, 6 | 891 | |||
AK316304.1 | - | 5, 6 | 851 | |||
AK316506.1 | - | 5, 6 | 719 | |||
BC014519.1 | - | 5, 6 | 753 | 3842039 | ||
BC068561.1 | - | 5, 6 | 786 | 5259336 | ||
BQ685792.1 | - | 5, 6 | 814 |