Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321357.1 | NP_001308286.1 | 14 | 1996 | ||
NM_001321358.1 | NP_001308287.1 | 14 | 1393 | |||
NM_001321359.1 | NP_001308288.1 | 14 | 1388 | |||
NM_001321361.1 | NP_001308290.1 | 14 | 1390 | |||
NM_001321362.1 | NP_001308291.1 | 13 | 1366 | |||
NM_001321363.1 | NP_001308292.1 | 13 | 1273 | |||
NM_013392.3 | NP_037524.1 | 13 | 1364 | |||
GenBank mRNA | AF113249.1 | - | 13 | 1276 | ||
AK001946.1 | - | 13 | 1254 | |||
AK027538.1 | - | 13 | 1272 | |||
AK092860.1 | - | 6 | 1674 | |||
AK122664.1 | - | 14 | 1996 | |||
AK130619.1 | - | 6 | 408 | |||
AK223136.1 | - | 13 | 1260 | |||
AK301345.1 | - | 14 | 1207 | |||
AL136682.1 | - | 13 | 1251 | |||
BC001221.1 | - | 13 | 1278 | 3534543 |