Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCHIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197107.1 | NP_001184036.1 | 5 | 1568 | ||
NM_001197108.1 | NP_001184037.1 | 4 | 911 | |||
NM_001197109.1 | NP_001184038.1 | 4 | 628 | |||
NM_014575.3 | NP_055390.1 | 5 | 1607 | |||
GenBank mRNA | AF070614.1 | - | 4 | 717 | ||
AF145713.1 | - | 5 | 1102 | |||
AF145714.1 | - | 4 | 406 | |||
AF145715.1 | - | 5 | 1063 | |||
AK091871.1 | - | 4 | 549 | |||
AK309223.1 | - | 5 | 1015 | |||
AK314662.1 | - | 5 | 1550 | |||
BC005947.1 | - | 4 | 444 | 4246326 | ||
BC036535.2 | - | 5 | 1607 | 5263291 | ||
BX648179.1 | - | 3 | 2125 |
Gene Symbol: IQCJ-SCHIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197113.1 | NP_001184042.1 | 8 | 1442 | ||
NM_001197114.1 | NP_001184043.1 | 7 | 1361 | |||
GenBank mRNA | AK096479.1 | - | 8 | 1348 | ||
DQ157847.1 | - | 6 | 1180 | |||
DQ157848.1 | - | 7 | 1261 |