Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CD44 SIRNA Assays ›
Gene Symbol:
CD44
Gene Name:
CD44 molecule (Indian blood group)
Gene Aliases:
CDW44, CSPG8, ECMR-III, HCELL, HUTCH-I, IN, LHR, MC56, MDU2, MDU3, MIC4, Pgp1
Chromosome Location:
Chr.11: 35138870 - 35232402 on Build GRCh38
UniGene ID:
Hs.502328
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s2683
Validated
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CD44
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000610.3
NP_000601.3
2
514
NM_001001389.1
NP_001001389.1
2
514
NM_001001390.1
NP_001001390.1
2
514
NM_001001391.1
NP_001001391.1
2
514
NM_001001392.1
NP_001001392.1
2
514
NM_001202555.1
NP_001189484.1
2
514
NM_001202556.1
NP_001189485.1
2
514
NM_001202557.1
NP_001189486.1
2
514
XM_005253231.2
XP_005253288.1
2
241
XM_005253232.2
XP_005253289.1
2
241
XM_005253233.2
XP_005253290.1
2
241
XM_005253234.2
XP_005253291.1
2
241
XM_005253235.2
XP_005253292.1
2
241
XM_005253238.2
XP_005253295.1
2
241
XM_005253239.2
XP_005253296.1
2
241
XM_005253240.2
XP_005253297.1
2
241
XM_006718388.1
XP_006718451.1
2
241
XM_006718390.3
XP_006718453.1
2
241
XM_011520482.1
XP_011518784.1
2
241
XM_011520483.1
XP_011518785.1
2
241
XM_011520484.1
XP_011518786.1
2
241
XM_011520485.1
XP_011518787.1
2
241
XM_011520486.1
XP_011518788.1
2
241
XM_011520487.2
XP_011518789.1
2
241
XM_011520488.1
XP_011518790.1
2
241
XM_011520489.2
XP_011518791.1
2
241
XM_017018583.1
XP_016874072.1
2
241
XM_017018584.1
XP_016874073.1
2
241
XM_017018585.1
XP_016874074.1
2
241
GenBank mRNA
AF098641.1
-
2
98
AJ251595.1
-
2
258
AK290424.1
-
2
80
AK297776.1
-
2
216
AL832642.2
-
2
514
AY101192.1
-
2
196
AY101193.1
-
2
197
BC004372.1
-
2
211
3638681
BC052287.1
-
2
202
BC067348.1
-
2
197
EF581837.1
-
2
91
FJ216964.1
-
2
80
M24915.1
-
2
195
M25078.1
-
2
78
M59040.1
-
2
108
S66400.1
-
2
130
X55150.1
-
2
203
X56794.1
-
2
195
X66733.1
-
2
209
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00174139_m1
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
cell-matrix adhesion
single organismal cell-cell adhesion
extracellular matrix disassembly
extracellular matrix organization
hyaluronan catabolic process
positive regulation of peptidyl-serine phosphorylation
positive regulation of heterotypic cell-cell adhesion
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
negative regulation of DNA damage response, signal transduction by p53 class mediator
cellular response to fibroblast growth factor stimulus
positive regulation of peptidyl-tyrosine phosphorylation
leukocyte migration
cartilage development
interferon-gamma-mediated signaling pathway
positive regulation of ERK1 and ERK2 cascade
monocyte aggregation
positive regulation of monocyte aggregation
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
cytokine receptor activity
protein binding
collagen binding
hyaluronic acid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-6c48f4b768-sx5kz:80/10.239.163.240:80.
git-commit: a2f5e26549390d543f5d2d1fbad8092df65fbda4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.10.0-2024.06.23-1.0