Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBQLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013438.4 | NP_038466.2 | 8, 9 | 1917 | ||
NM_053067.2 | NP_444295.1 | 7, 8 | 1833 | |||
GenBank mRNA | AB035275.1 | - | 8, 9 | 1513 | ||
AB209468.1 | - | 6, 7 | 3811 | |||
AF176069.3 | - | 8, 9 | 1343 | |||
AF293384.1 | - | 8, 9 | 1343 | |||
AK027556.1 | - | 8, 9 | 1616 | |||
AL136643.1 | - | 8, 9 | 1621 | |||
BC010066.2 | - | 7, 8 | 1513 | 4110398 | ||
BC039294.1 | - | 8, 9 | 1608 | 5263680 |