Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271711.1 | NP_001258640.1 | 4 | 532 | ||
NM_001271712.1 | NP_001258641.1 | 2 | 162 | |||
NM_014580.4 | NP_055395.2 | 4 | 532 | |||
NR_073416.1 | 2 | 162 | ||||
XM_006717084.3 | XP_006717147.1 | 4 | 527 | |||
XM_011518602.2 | XP_011516904.1 | 4 | 526 | |||
XM_011518603.2 | XP_011516905.1 | 2 | 187 | |||
XM_011518604.2 | XP_011516906.1 | 2 | 189 | |||
XM_017014653.1 | XP_016870142.1 | 2 | 152 | |||
GenBank mRNA | AF289587.1 | - | 4 | 439 | ||
AJ245937.1 | - | 4 | 442 | |||
BC019043.2 | - | 4 | 484 | 4641145 | ||
BM044230.1 | - | 2 | 63 | |||
BX395379.2 | - | 2 | 87 | |||
HY025817.1 | - | 4 | 397 | |||
Y17801.1 | - | 4 | 465 |