Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OBP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288987.1 | NP_001275916.1 | 2, 3 | 253 | ||
NM_014581.3 | NP_055396.1 | 2, 3 | 253 | |||
NR_110242.1 | 2, 3 | 253 | ||||
XM_006717086.3 | XP_006717149.1 | 2, 3 | 321 | |||
XM_017014654.1 | XP_016870143.1 | 3, 4 | 489 | |||
XM_017014655.1 | XP_016870144.1 | 4, 5 | 700 | |||
GenBank mRNA | AJ251026.1 | - | 2, 3 | 244 | ||
AJ251027.1 | - | 2, 3 | 244 | |||
AY358981.1 | - | 2, 3 | 246 | |||
BC096717.1 | - | 2, 3 | 228 | 40003080 | ||
BC098125.1 | - | 2, 3 | 228 | 40003075 | ||
BC098151.1 | - | 2, 3 | 228 | 40003079 | ||
BC098340.1 | - | 2, 3 | 228 | 40003073 | ||
HY046406.1 | - | 2, 3 | 253 |
Gene Symbol: OBP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293189.1 | NP_001280118.1 | 2, 3 | 244 | ||
NM_014582.2 | NP_055397.1 | 2, 3 | 244 | |||
NR_120603.1 | 2, 3 | 244 | ||||
XM_017014656.1 | XP_016870145.1 | 3, 4 | 1120 | |||
GenBank mRNA | AJ251021.1 | - | 2, 3 | 244 | ||
AJ251022.1 | - | 2, 3 | 244 | |||
AJ251024.1 | - | 2, 3 | 244 | |||
AM087898.1 | - | 2, 3 | 160 | |||
BC069563.1 | - | 2, 3 | 229 | 7262146 | ||
BC143803.1 | - | 2, 3 | 230 | 9052317 |