Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAG3L5P-PVRIG2P-PILRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036569.1 | 18 | 3449 | |||
NR_036570.1 | 17 | 2773 | ||||
GenBank mRNA | AF217981.1 | - | 9 | 2178 | ||
AJ400846.1 | - | 7 | 1289 | |||
AK056412.1 | - | 18 | 3400 | |||
AK298676.1 | - | 5 | 1239 | |||
AL834317.1 | - | 17 | 2595 | |||
AL834336.1 | - | 5 | 1118 |
Gene Symbol: PILRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178238.3 | NP_839956.1 | 4 | 1204 | ||
GenBank mRNA | AJ400845.1 | - | 4 | 1209 | ||
BC050547.1 | - | 7 | 1251 | 5755993 |