Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OBP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293189.1 | NP_001280118.1 | 5 | 546 | ||
NM_001293193.1 | NP_001280122.1 | 4 | 412 | |||
NM_014582.2 | NP_055397.1 | 5 | 481 | |||
NR_120603.1 | 5 | 530 | ||||
XM_017014656.1 | XP_016870145.1 | 6 | 1422 | |||
GenBank mRNA | AJ251021.1 | - | 5 | 481 | ||
AJ251022.1 | - | 5 | 530 | |||
AJ251024.1 | - | 5 | 546 | |||
BC069563.1 | - | 5 | 466 | 7262146 | ||
BC143803.1 | - | 5 | 532 | 9052317 |