Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PACSIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199583.2 | NP_001186512.1 | 3 | 377 | ||
NM_020804.4 | NP_065855.1 | 3 | 390 | |||
XM_011514541.1 | XP_011512843.1 | 3 | 245 | |||
GenBank mRNA | AB037800.2 | - | 3 | 347 | ||
AF242529.1 | - | 3 | 313 | |||
AK289956.1 | - | 3 | 375 | |||
AK294132.1 | - | 3 | 377 | |||
AL834211.1 | - | 3 | 323 | |||
BC040228.1 | - | 3 | 362 | 5173129 | ||
BP211882.1 | - | 3 | 390 | |||
BP251080.1 | - | 3 | 377 | |||
DC312410.1 | - | 3 | 377 |