Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BAZ2B SIRNA Assays ›
Gene Symbol:
BAZ2B
Gene Name:
bromodomain adjacent to zinc finger domain 2B
Gene Aliases:
WALp4
Chromosome Location:
Chr.2: 159315312 - 159712439 on Build GRCh38
UniGene ID:
Hs.470369
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s26867
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BAZ2B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289975.1
NP_001276904.1
31
5881
NM_013450.3
NP_038478.2
32
5989
XM_005246488.2
XP_005246545.2
33
5865
XM_005246489.4
XP_005246546.2
32
6968
XM_005246492.4
XP_005246549.2
32
6963
XM_006712461.3
XP_006712524.2
36
6439
XM_011511038.2
XP_011509340.1
33
5963
XM_011511039.1
XP_011509341.1
30
5586
XM_011511042.1
XP_011509344.1
31
5619
XM_011511043.1
XP_011509345.1
29
5484
XM_011511044.1
XP_011509346.1
30
5511
XM_011511045.1
XP_011509347.1
30
5424
XM_011511047.1
XP_011509349.1
32
5771
XM_011511048.2
XP_011509350.1
32
5836
XM_011511050.1
XP_011509352.1
30
5319
XM_011511051.1
XP_011509353.1
30
5505
XM_011511052.1
XP_011509354.1
29
5223
XM_011511053.2
XP_011509355.1
31
5619
XM_011511054.2
XP_011509356.1
31
5619
XM_011511056.1
XP_011509358.1
21
3239
XM_017003918.1
XP_016859407.1
36
6429
XM_017003919.1
XP_016859408.1
36
6423
XM_017003920.1
XP_016859409.1
38
6640
XM_017003921.1
XP_016859410.1
33
5963
XM_017003922.1
XP_016859411.1
33
5859
XM_017003923.1
XP_016859412.1
31
5613
XM_017003924.1
XP_016859413.1
30
5511
XM_017003925.1
XP_016859414.1
32
5867
XM_017003926.1
XP_016859415.1
32
5725
XM_017003927.1
XP_016859416.1
32
5765
XM_017003928.1
XP_016859417.1
32
5675
XM_017003929.1
XP_016859418.1
32
5757
XM_017003930.1
XP_016859419.1
32
5565
XM_017003931.1
XP_016859420.1
32
5663
XM_017003932.1
XP_016859421.1
29
5316
XM_017003933.1
XP_016859422.1
31
5542
XM_017003934.1
XP_016859423.1
31
5437
XM_017003935.1
XP_016859424.1
30
5375
XM_017003936.1
XP_016859425.1
30
5329
XM_017003937.1
XP_016859426.1
25
4036
GenBank mRNA
AB032255.1
-
27
5362
AB040909.2
-
31
5849
AK074895.1
-
10
2107
AL080173.1
-
3
429
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-69b65fdbbd-mdpxp:80/100.66.77.49:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline