Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001778.3 | NP_001769.2 | 4 | 760 | ||
XM_005245625.1 | XP_005245682.1 | 3 | 464 | |||
XM_011510171.1 | XP_011508473.1 | 5 | 828 | |||
XM_017002867.1 | XP_016858356.1 | 3 | 444 | |||
GenBank mRNA | BC016182.1 | - | 4 | 704 | 4850933 | |
BT019813.1 | - | 4 | 672 | |||
CR457012.1 | - | 4 | 672 | |||
M37766.1 | - | 4 | 692 | |||
M59904.1 | - | 4 | 689 | |||
X06341.1 | - | 4 | 708 |