Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERO1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014584.2 | NP_055399.1 | 12 | 1311 | ||
XM_011536684.2 | XP_011534986.1 | 11 | 1118 | |||
GenBank mRNA | AF081886.1 | - | 12 | 1087 | ||
AF123887.1 | - | 10 | 613 | |||
AK292839.1 | - | 12 | 909 | |||
AY358463.1 | - | 12 | 1033 | |||
BC008674.1 | - | 12 | 1083 | 3866930 | ||
BC012941.1 | - | 12 | 1094 | 3868538 |