Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021025.3 | NP_066305.2 | 3 | 1271 | ||
GenBank mRNA | BC017291.2 | - | 3 | 1271 | 4906239 |